Assay Details
| Gene Symbol: SEC11C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103798.2 | XP_014959284.2 | 4 - 5 | 565 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC11C| Gene Symbol: SEC11C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103798.2 | XP_014959284.2 | 4 - 5 | 565 | 80 | |
Set Membership: |
|