Assay Details
| Gene Symbol: ECT2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182978.2 | XP_012038368.1 | 17 - 18 | 2349 | 73 | |
| XM_027972496.1 | XP_027828297.1 | 18 - 19 | 2462 | 73 | ||
| XM_027972497.1 | XP_027828298.1 | 17 - 18 | 2333 | 73 | ||
More Information
Set Membership: |
|

ECT2L
Genomic Map