Assay Details
| Gene Symbol: CX3CL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100450.2 | XP_014955936.2 | 2 - 3 | 336 | 65 | |
| XM_027977869.1 | XP_027833670.1 | 2 - 3 | 366 | 65 | ||
More Information
Set Membership: |
|

CX3CL1
Genomic Map