Assay Details
| Gene Symbol: BMF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096939.2 | XP_014952425.1 | 4 - 5 | 625 | 61 | |
| XM_027971669.1 | XP_027827470.1 | 4 - 5 | 1278 | 61 | ||
More Information
Set Membership: |
|

BMF
Genomic Map