Assay Details
| Gene Symbol: IKBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006635.4 | XP_004006684.2 | 1 - 2 | 445 | 79 | |
| XM_027967646.1 | XP_027823447.1 | 1 - 2 | 416 | 79 | ||
More Information
Set Membership: |
|

IKBIP
Genomic Map