Assay Details
| Gene Symbol: TOR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007147.3 | XP_004007196.3 | 2 - 3 | 698 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOR1A| Gene Symbol: TOR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007147.3 | XP_004007196.3 | 2 - 3 | 698 | 67 | |
Set Membership: |
|