Assay Details
| Gene Symbol: SERPINB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020573.4 | XP_004020622.1 | 6 - 7 | 840 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERPINB5| Gene Symbol: SERPINB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020573.4 | XP_004020622.1 | 6 - 7 | 840 | 69 | |
Set Membership: |
|