Assay Details
| Gene Symbol: C17H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980242.1 | XP_027836043.1 | 2 - 3 | 288 | 85 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C17H4orf45| Gene Symbol: C17H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980242.1 | XP_027836043.1 | 2 - 3 | 288 | 85 | |