Assay Details
| Gene Symbol: ALPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012152818.3 | XP_012008208.2 | 10 - 11 | 1405 | 60 | |
| XM_012152824.3 | XP_012008214.2 | 10 - 11 | 1409 | 60 | ||
| XM_012152832.3 | XP_012008222.2 | 10 - 11 | 1358 | 60 | ||
| XM_027965561.1 | XP_027821362.1 | 10 - 11 | 1379 | 60 | ||
More Information
Set Membership: |
|

ALPL
Genomic Map