Assay Details
| Gene Symbol: CCND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959928.1 | XP_027815729.1 | 3 - 4 | 790 | 70 | |
| GenBank mRNA | HQ825087.1 | - | 2 - 3 | 340 | 70 | |
More Information
Set Membership: |
|

CCND1
Genomic Map