Assay Details
| Gene Symbol: SLC22A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183210.3 | XP_012038600.3 | 7 - 8 | 1295 | 60 | |
| XM_027972635.1 | XP_027828436.1 | 7 - 8 | 1295 | 60 | ||
| GenBank mRNA | HQ667515.1 | - | 7 - 8 | 947 | 60 | |
More Information
Set Membership: |
|

SLC22A3
Genomic Map