Assay Details
| Gene Symbol: CEP85L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097328.2 | XP_014952814.2 | 2 - 3 | 248 | 70 | |
| XM_027972343.1 | XP_027828144.1 | 2 - 3 | 248 | 70 | ||
More Information
Set Membership: |
|

CEP85L
Genomic Map