Assay Details
| Gene Symbol: TBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097549.2 | XP_014953035.2 | 4 - 5 | 850 | 66 | |
| XM_027972700.1 | XP_027828501.1 | 4 - 5 | 847 | 66 | ||
| GenBank mRNA | AY954925.1 | - | 1 - 2 | 82 | 66 | |
More Information
Set Membership: |
|

TBP
Genomic Map