Assay Details
| Gene Symbol: LOC101117577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965980.1 | XP_027821781.1 | 19 - 20 | 2263 | 101 | |
| Gene Symbol: LOC114113811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965979.1 | XP_027821780.1 | 8 - 9 | 931 | 101 | |
More Information
Set Membership: |
|

LOC101117577
Genomic Map