Assay Details
| Gene Symbol: MAPK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177604.3 | XP_012032994.1 | 2 - 3 | 186 | 98 | |
| XM_027969268.1 | XP_027825069.1 | 2 - 3 | 282 | 98 | ||
| XM_027969269.1 | XP_027825070.1 | 2 - 3 | 282 | 98 | ||
| XM_027969270.1 | XP_027825071.1 | 2 - 3 | 282 | 98 | ||
| XM_027969271.1 | XP_027825072.1 | 2 - 3 | 284 | 98 | ||
| XM_027969272.1 | XP_027825073.1 | 2 - 3 | 288 | 98 | ||
More Information
Set Membership: |
|

MAPK9
Genomic Map