Assay Details
| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958489.1 | XP_027814290.1 | 1 - 2 | 1078 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAD2L1BP| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958489.1 | XP_027814290.1 | 1 - 2 | 1078 | 107 | |
Set Membership: |
|