Assay Details
| Gene Symbol: LOC101120448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019111.4 | XP_004019160.2 | 3 - 4 | 335 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120448| Gene Symbol: LOC101120448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019111.4 | XP_004019160.2 | 3 - 4 | 335 | 97 | |
Set Membership: |
|