Assay Details
| Gene Symbol: LOC106990117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958887.1 | XP_027814688.1 | 7 - 8 | 1271 | 72 | |
| XM_027958888.1 | XP_027814689.1 | 8 - 9 | 1700 | 72 | ||
| XM_027958889.1 | XP_027814690.1 | 7 - 8 | 1226 | 72 | ||
| XM_027958890.1 | XP_027814691.1 | 7 - 8 | 1192 | 72 | ||
| XM_027958891.1 | XP_027814692.1 | 6 - 7 | 994 | 72 | ||
More Information
Set Membership: |
|

LOC106990117
Genomic Map