Assay Details
| Gene Symbol: DGCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956940.1 | XP_027812741.1 | 12 - 13 | 2425 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DGCR8| Gene Symbol: DGCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956940.1 | XP_027812741.1 | 12 - 13 | 2425 | 56 | |
Set Membership: |
|