Assay Details
| Gene Symbol: TNFRSF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104084.2 | XP_014959570.1 | 2 - 3 | 354 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF17| Gene Symbol: TNFRSF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104084.2 | XP_014959570.1 | 2 - 3 | 354 | 70 | |
Set Membership: |
|