Assay Details
| Gene Symbol: LOC101117326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008067.4 | XP_004008116.1 | 7 - 8 | 796 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117326| Gene Symbol: LOC101117326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008067.4 | XP_004008116.1 | 7 - 8 | 796 | 100 | |
Set Membership: |
|