Assay Details
| Gene Symbol: MAP2K4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974871.1 | XP_027830672.1 | 8 - 9 | 891 | 67 | |
| XM_027974872.1 | XP_027830673.1 | 9 - 10 | 924 | 67 | ||
More Information
Set Membership: |
|

MAP2K4
Genomic Map