Assay Details
| Gene Symbol: GLRX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017981.4 | XP_004018030.1 | 1 - 2 | 312 | 72 | |
| Gene Symbol: LOC114110487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961092.1 | XP_027816893.1 | 1 | 329 | 72 | |
More Information
Set Membership: |
|

GLRX5
Genomic Map