Assay Details
| Gene Symbol: LOC101115913 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018506.4 | XP_004018555.1 | 5 - 6 | 713 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101115913| Gene Symbol: LOC101115913 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018506.4 | XP_004018555.1 | 5 - 6 | 713 | 89 | |
Set Membership: |
|