Assay Details
| Gene Symbol: LOC101121723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019389.4 | XP_004019438.2 | 3 - 4 | 541 | 87 | |
| XM_012101586.3 | XP_011956976.2 | 3 - 4 | 520 | 87 | ||
More Information
Set Membership: |
|

LOC101121723
Genomic Map