Assay Details
| Gene Symbol: LOC101121723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019389.4 | XP_004019438.2 | 9 - 10 | 1241 | 101 | |
| XM_012101586.3 | XP_011956976.2 | 9 - 10 | 1232 | 101 | ||
More Information
Set Membership: |
|

LOC101121723
Genomic Map