Assay Details
| Gene Symbol: ZNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023243.4 | XP_004023292.3 | 3 - 4 | 507 | 76 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF41| Gene Symbol: ZNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023243.4 | XP_004023292.3 | 3 - 4 | 507 | 76 | |