Assay Details
| Gene Symbol: LOC101115486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098423.3 | XP_011953813.2 | 18 - 19 | 5192 | 96 | |
| XM_012098428.3 | XP_011953818.2 | 13 - 14 | 3176 | 96 | ||
| XM_027975368.1 | XP_027831169.1 | 22 - 23 | 5899 | 96 | ||
| XM_027975370.1 | XP_027831171.1 | 22 - 23 | 5899 | 96 | ||
| XM_027975372.1 | XP_027831173.1 | 21 - 22 | 5833 | 96 | ||
| XM_027975373.1 | XP_027831174.1 | 20 - 21 | 5779 | 96 | ||

LOC101115486
Genomic Map