Assay Details
| Gene Symbol: LOC101115486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098423.3 | XP_011953813.2 | 28 - 29 | 6779 | 65 | |
| XM_012098428.3 | XP_011953818.2 | 23 - 24 | 4763 | 65 | ||
| XM_027975368.1 | XP_027831169.1 | 32 - 33 | 7486 | 65 | ||
| XM_027975369.1 | XP_027831170.1 | 32 - 33 | 7483 | 65 | ||
| XM_027975370.1 | XP_027831171.1 | 32 - 33 | 7486 | 65 | ||
| XM_027975371.1 | XP_027831172.1 | 32 - 33 | 7480 | 65 | ||
| XM_027975372.1 | XP_027831173.1 | 31 - 32 | 7420 | 65 | ||
| XM_027975373.1 | XP_027831174.1 | 30 - 31 | 7366 | 65 | ||

LOC101115486
Genomic Map