Assay Details
| Gene Symbol: LOC101115486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098423.3 | XP_011953813.2 | 8 - 9 | 4085 | 102 | |
| XM_027975368.1 | XP_027831169.1 | 12 - 13 | 4792 | 102 | ||
| XM_027975369.1 | XP_027831170.1 | 12 - 13 | 4792 | 102 | ||
| XM_027975370.1 | XP_027831171.1 | 12 - 13 | 4792 | 102 | ||
| XM_027975371.1 | XP_027831172.1 | 12 - 13 | 4792 | 102 | ||
More Information
Set Membership: |
|

LOC101115486
Genomic Map