Assay Details
| Gene Symbol: LOC101115486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098423.3 | XP_011953813.2 | 10 - 11 | 4344 | 86 | |
| XM_012098428.3 | XP_011953818.2 | 5 - 6 | 2328 | 86 | ||
| XM_027975368.1 | XP_027831169.1 | 14 - 15 | 5051 | 86 | ||
| XM_027975369.1 | XP_027831170.1 | 14 - 15 | 5051 | 86 | ||
| XM_027975370.1 | XP_027831171.1 | 14 - 15 | 5051 | 86 | ||
| XM_027975371.1 | XP_027831172.1 | 14 - 15 | 5051 | 86 | ||
| XM_027975372.1 | XP_027831173.1 | 13 - 14 | 4985 | 86 | ||
| XM_027975373.1 | XP_027831174.1 | 12 - 13 | 4931 | 86 | ||

LOC101115486
Genomic Map