Assay Details
| Gene Symbol: LOC101116577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182163.3 | XP_012037553.1 | 2 - 3 | 1203 | 84 | |
| XM_027972087.1 | XP_027827888.1 | 2 - 3 | 1203 | 84 | ||
| XM_027972088.1 | XP_027827889.1 | 11 - 12 | 3574 | 84 | ||
| XM_027972089.1 | XP_027827890.1 | 2 - 3 | 1203 | 84 | ||

LOC101116577
Genomic Map