Assay Details
| Gene Symbol: STK40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015091863.2 | XP_014947349.2 | 9 - 10 | 1202 | 58 | |
| XM_027967921.1 | XP_027823722.1 | 9 - 10 | 1445 | 58 | ||
| XM_027967927.1 | XP_027823728.1 | 9 - 10 | 1271 | 58 | ||
More Information
Set Membership: |
|

STK40
Genomic Map