Assay Details
| Gene Symbol: LOC101120993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012148264.3 | XP_012003654.2 | 8 - 9 | 908 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120993| Gene Symbol: LOC101120993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012148264.3 | XP_012003654.2 | 8 - 9 | 908 | 79 | |