Assay Details
| Gene Symbol: LOC101118866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967501.1 | XP_027823302.1 | 5 - 6 | 670 | 120 | |
| XM_027967502.1 | XP_027823303.1 | 3 - 4 | 924 | 120 | ||
| XM_027967503.1 | XP_027823304.1 | 4 - 5 | 310 | 120 | ||
| XM_027967504.1 | XP_027823305.1 | 4 - 5 | 485 | 120 | ||
More Information
Set Membership: |
|

LOC101118866
Genomic Map