Assay Details
| Gene Symbol: LTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102797.2 | XP_014958283.1 | 1 - 2 | 110 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102797.2 | XP_014958283.1 | 1 - 2 | 110 | 90 | |