Assay Details
| Gene Symbol: YTHDF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972989.1 | XP_027828790.1 | 7 - 8 | 2412 | 147 | |
| XM_027972990.1 | XP_027828791.1 | 6 - 7 | 2266 | 147 | ||
| XM_027972991.1 | XP_027828792.1 | 6 - 7 | 2367 | 147 | ||
| XM_027972992.1 | XP_027828793.1 | 5 - 6 | 2221 | 147 | ||
More Information
Set Membership: |
|

YTHDF3
Genomic Map