Assay Details
| Gene Symbol: LOC101117325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968051.1 | XP_027823852.1 | 4 - 5 | 546 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117325| Gene Symbol: LOC101117325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968051.1 | XP_027823852.1 | 4 - 5 | 546 | 103 | |
Set Membership: |
|