Assay Details
| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003064.4 | XP_004003113.1 | 2 - 3 | 1061 | 94 | |
| XM_027957510.1 | XP_027813311.1 | 1 - 2 | 4789 | 94 | ||
| XM_027957512.1 | XP_027813313.1 | 2 - 3 | 485 | 94 | ||
More Information
Set Membership: |
|

CRYGS
Genomic Map