Assay Details
| Gene Symbol: LOC101102057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008561.4 | XP_004008610.1 | 4 - 5 | 371 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101102057| Gene Symbol: LOC101102057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008561.4 | XP_004008610.1 | 4 - 5 | 371 | 101 | |
Set Membership: |
|