Assay Details
| Gene Symbol: DKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009640.4 | XP_004009689.2 | 3 - 4 | 835 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009640.4 | XP_004009689.2 | 3 - 4 | 835 | 81 | |