Assay Details
| Gene Symbol: LOC114115243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970755.1 | XP_027826556.1 | 2 - 3 | 161 | 75 | |
| Gene Symbol: LOC494440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970756.1 | XP_027826557.1 | 20 - 21 | 2916 | 75 | |
More Information
Set Membership: |
|

LOC114115243
Genomic Map