Assay Details
| Gene Symbol: LOC114118747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980145.1 | XP_027835946.1 | 1 | 1829 | 82 | |
| Gene Symbol: NLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975242.1 | XP_027831043.1 | 5 - 6 | 2487 | 82 | |
More Information
Set Membership: |
|

LOC114118747
Genomic Map