Assay Details
| Gene Symbol: ADRA1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014355.3 | XP_004014404.2 | 1 - 2 | 1311 | 64 | |
| GenBank mRNA | EU723258.1 | - | 1 - 2 | 323 | 64 | |
More Information
Set Membership: |
|

ADRA1D
Genomic Map