Assay Details
| Gene Symbol: CIAPIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015023.4 | XP_004015072.3 | 8 - 9 | 921 | 57 | |
| XM_012189824.3 | XP_012045214.3 | 8 - 9 | 924 | 57 | ||
More Information
Set Membership: |
|

CIAPIN1
Genomic Map