Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018197.4 | XP_004018246.1 | 4 - 5 | 932 | 78 | |
| XM_012099295.3 | XP_011954685.1 | 4 - 5 | 932 | 78 | ||
More Information
Set Membership: |
|

AZI2
Genomic Map