Assay Details
| Gene Symbol: BTNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958971.1 | XP_027814772.1 | 2 - 3 | 424 | 88 | |
| XM_027958972.1 | XP_027814773.1 | 2 - 3 | 424 | 88 | ||
| GenBank mRNA | JF735992.1 | - | 1 - 2 | 342 | 88 | |
More Information
Set Membership: |
|

BTNL2
Genomic Map