Assay Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972090.1 | XP_027827891.1 | 1 - 2 | 271 | 81 | |
| XM_027972091.1 | XP_027827892.1 | 1 - 2 | 302 | 81 | ||
| GenBank mRNA | GQ468498.1 | - | 1 - 2 | 215 | 81 | |
More Information
Set Membership: |
|

DIO2
Genomic Map