Assay Details
| Gene Symbol: CCDC157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980269.1 | XP_027836070.1 | 5 - 6 | 1318 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC157| Gene Symbol: CCDC157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980269.1 | XP_027836070.1 | 5 - 6 | 1318 | 58 | |
Set Membership: |
|