Assay Details
| Gene Symbol: LOC101103238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971406.1 | XP_027827207.1 | 3 - 4 | 464 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103238| Gene Symbol: LOC101103238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971406.1 | XP_027827207.1 | 3 - 4 | 464 | 83 | |
Set Membership: |
|